missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human MCP-1 Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human MCP-1.

Brand:  Invitrogen™ BMS281TENCE

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15581877

  • £2180.00 / Pack of 10


Estimated Shipment date
from Supplier 26-02-2020
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human MCP-1
HRP-Conjugate anti-human MCP-1 monoclonal antibody
Human MCP-1 Standard lyophilized, 2ng/mL upon reconstitution
Control high, lyophilized
Control low, lyophilized
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

HRP
2.3pg/mL
CCL2 (MCP-1)
10 x 96 Tests
Serum
ELISA
20 μL
Monocyte chemoattractant protein 1, MCAF, HC11
Human
Videos
Documents

For Research Use Only