missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human M-CSF, Recombinant Protein, eBioscience™
Recombinant Protein
Brand: Affymetrix eBioscience 14-8789-62
Code : NEW
Additional Details : Weight : 0.09500kg
Description
M-CSF (Macrophage colony-stimulating factor, CSF-1) is a survival factor essential for the proliferation and development of monocytes, macrophages, and osteoclast progenitor cells. M-CSF also induces VEGF (vascular endothelial growth factor) secretion by macrophages, thereby mediating mobilization of endothelial progenitor cells and neovascularization. M-CSF is present as several bioactive isoforms that differ in potency and stability. The full-length protein is synthesized as a membrane-spanning protein that can be expressed on the cell surface or further cleaved and modified in the secretory vesicle. Further, M-CSF is a disulfide-bonded homodimer which is processed into one of two isoforms, a glycoprotein or a proteoglycan that has been modified by the addition of chondroitin sulfate to each subunit. Binding of M-CSF to its receptor, c-Fms (CSF-1R or CD115) induces dimerization of the receptor followed by internalization and degradation of the complex. Functionally, M-CSF is known to stimulate differentiation of hematopoietic stem cells to monocyte-macrophage cell populations in culture. M-CSF acts through the CSF receptor 1. Although human M-CSF shows activity on mouse cells, mouse CSF shows no activity on human cells. Description: Human Macrophage Colony Stimulating Factor (M-CSF, also called CSF-1) is a ∽36 kDa disulfide-linked homodimer produced by a variety of cells including fibroblasts, macrophages, bone marrow stromal cells and endothelial cells. M-CSF regulates macrophage survival, proliferation and differentiation. M-CSF also enhances macrophage killing of tumor cells and microorganisms and regulates cytokine production from macrophages. Applications Reported: Recombinant human M-CSF is biologically active and can promote proliferation of M-NFS-60 cells in culture. Applications Tested: The bioactivity of this protein, as determined in a cell proliferation assay on M-NFS-60 cells, is 3ng/mL. This corresponds to a specific activity of 3.3 X 10e5 Units/mg. Source: Human M-CSF Glu34-Ser190 accession &Num; NM-172211 expressed in insect cells with a C-terminal 6X His tag. Bioactivity: The bioactivity of this protein, as determined in a cell proliferation assay on M-NFS-60 cells, is 3ng/mL. This corresponds to a specific activity of 3.3 X 10e5 Units/mg. Endotoxin: Less than 0.01ng/ug cytokine as determined by the LAL assay. Purity: >98% as determined by SDS-PAGE. Storage and handling: For best recovery, always quick-spin vial prior to opening. For dilution of current stock, always include carrier protein (1% BSA or 10% FBS) in the buffered saline diluent. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2um post-manufacturing filtered.Specifications
P09603 | |
Control, Functional Assay | |
1435 | |
Human M-CSF | |
SDS-PAGE | |
Insect Cells | |
RUO | |
Less than 0.01ng/μg cytokine as determined by the LAL assay. | |
CSF1 | |
Recombinant Protein | |
Unconjugated | |
Baculovirus | |
Liquid | |
Affinity chromatography |
0.1mg/mL | |
PBS with 1% BSA and no preservative; pH 7.2 | |
36kDa | |
7.2 | |
5μg | |
-80°C | |
Human M-CSF Glu34-Ser190 accession # NM-172211 expressed in insect cells with a C-terminal 6X His tag. | |
Human M-CSF | |
The bioactivity of this protein, as determined in a cell proliferation assay on M-NFS-60 cells, is 3ng/mL. This corresponds to a specific activity of 3.3 X 105 Units/mg. | |
Human | |
Hu | |
Recombinant | |
Recombinant |