missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human LAP (TGF beta 1) ELISA Ready-SET-Go!™ Kit

Specifically engineered for accurate and precise measurement of LAP. TGF beta is synthesized as a precursor that contains LAP at the N-terminus and mature TGF beta at the C-terminus

Brand:  Invitrogen™ 88-50390-77

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15510877

  • £404.91 / Pack of 20


EU Stock 2
Add to basket

Includes: Capture Antibody Concentrate
Detection Antibody Concentrate
Human LAP standard (lyophilized): 20ng/mL upon reconstitution
Phosphate Buffered Saline Concentrate (PBS)
Assay Buffer A Concentrate (PBS with 1% Tween 20 and 10% BSA)
Sample Diluent B
pre-titrated Streptavidin-HRP
Tetramethylbenzidine (TMB) Substrate Solution

Description

Description

LAP Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)
Specifications

Specifications

ELISA
156.3pg/mL
10μL
2° to 8°C
Latency Associated Peptide, Pro-TGF beta 1, LAP/TGF beta 1
20 x 96 tests
Cell culture supernatant, serum and plasma (heparin)
Human
LAP
Videos

For Research Use Only