Learn More
Abnova™ Human KLRB1 (NP_002249, 67 a.a. - 225 a.a.) Partial Recombinant Protein with His tag
Used for SDS-PAGE
Brand: Abnova™ P5859.100ug
Additional Details : Weight : 0.02000kg
Description
Natural killer (NK) cells are lymphocytes that mediate cytotoxicity and secrete cytokines after immune stimulation. Several genes of the C-type lectin superfamily, including the rodent NKRP1 family of glycoproteins, are expressed by NK cells and may be involved in the regulation of NK cell function. The KLRB1 protein contains an extracellular domain with several motifs characteristic of C-type lectins, a transmembrane domain, and a cytoplasmic domain. The KLRB1 protein is classified as a type II membrane protein because it has an external C terminus. [provided by RefSeq]
Sequence: MGSSHHHHHHSSGLVPRGSHMGSMQKSSIEKCSVDIQQSRNKTTERPGLLNCPIYWQQLREKCLLFSHTVNPWNNSLADCSTKESSLLLIRDKDELIHTQNLIRDKAILFWIGLNFSLSEKNWKWINGSFLNSNDLEIRGDAKENSCISISQTSVYSEYCSTEIRWICQKELTPVRNKVYPDSSpecifications
SDS-PAGE | |
3820 | |
KLRB1 (Human) Recombinant Protein | |
3 ug/lane in 15% SDS-PAGE Stained with Coomassie Blue. Due to the protein nature, dimmers and multimers may be observed. | |
MGSSHHHHHHSSGLVPRGSHMGSMQKSSIEKCSVDIQQSRNKTTERPGLLNCPIYWQQLREKCLLFSHTVNPWNNSLADCSTKESSLLLIRDKDELIHTQNLIRDKAILFWIGLNFSLSEKNWKWINGSFLNSNDLEIRGDAKENSCISISQTSVYSEYCSTEIRWICQKELTPVRNKVYPDS | |
RUO | |
KLRB1 | |
E. coli | |
His | |
Liquid |
Liquid | |
21kDa | |
Escherichia coli expression system | |
100 ug | |
Store at -20°C. For long term storage store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CD161/CLEC5B/MGC138614/NKR/NKR-P1/NKR-P1A/NKRP1A/hNKR-P1A | |
KLRB1 | |
Recombinant | |
Escherichia coli expression system | |
>85% by SDS - PAGE |