missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human KLK1 Full-length ORF (NP_002248.1, 1 a.a. - 262 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
£280.00 - £425.00
Specifications
Accession Number | NP_002248.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 3816 |
Molecular Weight (g/mol) | 55.3kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16179677
|
Abnova™
H00003816-P01.10UG |
10 ug |
£280.00
10µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16189677
|
Abnova™
H00003816-P01.25UG |
25 ug |
£425.00
25µg |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Kallikreins are a subgroup of serine proteases having diverse physiological functions. Growing evidence suggests that many kallikreins are implicated in carcinogenesis and some have potential as novel cancer and other disease biomarkers. This gene is one of the fifteen kallikrein subfamily members located in a cluster on chromosome 19. This protein is functionally conserved in its capacity to release the vasoactive peptide, Lys-bradykinin, from low molecular weight kininogen. [provided by RefSeq]
Sequence: MWFLVLCLALSLGGTGAAPPIQSRIVGGWECEQHSQPWQAALYHFSTFQCGGILVHRQWVLTAAHCISDNYQLWLGRHNLFDDENTAQFVHVSESFPHPGFNMSLLENHTRQADEDYSHDLMLLRLTEPADTITDAVKVVELPTEEPEVGSTCLASGWGSIEPENFSFPDDLQCVDLKILPNDECKKAHVQKVTDFMLCVGHLEGGKDTCVGDSGGPLMCDGVLQGVTSWGYVPCGTPNKPSVAVRVLSYVKWIEDTIAENSSpecifications
NP_002248.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
55.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
MWFLVLCLALSLGGTGAAPPIQSRIVGGWECEQHSQPWQAALYHFSTFQCGGILVHRQWVLTAAHCISDNYQLWLGRHNLFDDENTAQFVHVSESFPHPGFNMSLLENHTRQADEDYSHDLMLLRLTEPADTITDAVKVVELPTEEPEVGSTCLASGWGSIEPENFSFPDDLQCVDLKILPNDECKKAHVQKVTDFMLCVGHLEGGKDTCVGDSGGPLMCDGVLQGVTSWGYVPCGTPNKPSVAVRVLSYVKWIEDTIAENS | |
RUO | |
KLK1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
3816 | |
KLK1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KLKR/Klk6/hK1 | |
KLK1 | |
Recombinant | |
wheat germ expression system |