Learn More
Abnova™ Human KLF12 Full-length ORF (NP_009180.3, 1 a.a. - 402 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00011278-P01.2ug
Additional Details : Weight : 0.00010kg
Description
Activator protein-2 alpha (AP-2 alpha) is a developmentally-regulated transcription factor and important regulator of gene expression during vertebrate development and carcinogenesis. The protein encoded by this gene is a member of the Kruppel-like zinc finger protein family and can repress expression of the AP-2 alpha gene by binding to a specific site in the AP-2 alpha gene promoter. Repression by the encoded protein requires binding with a corepressor, CtBP1. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]
Sequence: MNIHMKRKTIKNINTFENRMLMLDGMPAVRVKTELLESEQGSPNVHNYPDMEAVPLLLNNVKGEPPEDSLSVDHFQTQTEPVDLSINKARTSPTAVSSSPVSMTASASSPSSTSTSSSSSSRLASSPTVITSVSSASSSSTVLTPGPLVASASGVGGQQFLHIIHPVPPSSPMNLQSNKLSHVHRIPVVVQSVPVVYTAVRSPGNVNNTIVVPLLEDGRGHGKAQMDPRGLSPRQSKSDSDDDDLPNVTLDSVNETGSTALSIARAVQEVHPSPVSRVRGNRMNNQKFPCSISPFSIESTRRQRRSESPDSRKRRIHRCDFEGCNKVYTKSSHLKAHRRTHTGEKPYKCTWEGCTWKFARSDELTRHYRKHTGVKPFKCADCDRSFSRSDHLALHRRRHMLVSpecifications
NP_009180.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
70.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
2 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AP-2rep/AP2REP/HSPC122 | |
KLF12 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
11278 | |
KLF12 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MNIHMKRKTIKNINTFENRMLMLDGMPAVRVKTELLESEQGSPNVHNYPDMEAVPLLLNNVKGEPPEDSLSVDHFQTQTEPVDLSINKARTSPTAVSSSPVSMTASASSPSSTSTSSSSSSRLASSPTVITSVSSASSSSTVLTPGPLVASASGVGGQQFLHIIHPVPPSSPMNLQSNKLSHVHRIPVVVQSVPVVYTAVRSPGNVNNTIVVPLLEDGRGHGKAQMDPRGLSPRQSKSDSDDDDLPNVTLDSVNETGSTALSIARAVQEVHPSPVSRVRGNRMNNQKFPCSISPFSIESTRRQRRSESPDSRKRRIHRCDFEGCNKVYTKSSHLKAHRRTHTGEKPYKCTWEGCTWKFARSDELTRHYRKHTGVKPFKCADCDRSFSRSDHLALHRRRHMLV | |
RUO | |
KLF12 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |