missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human KLF10 Partial ORF (AAH11538.1, 111 a.a. - 210 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00007071-Q02.10ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16094865

  • £280.00 / 10µg
Estimated Shipment: 22-05-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Sequence: LSDTAKPHIAAPFKEEEKSPVSAPKLPKAQATSVIRHTADAQLCNHQTCPMKAASILNYQNNSFRRRTHLNVEAARKNIPCAAVSPNRSKCERNTVADVD
Specifications

Specifications

AAH11538.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
LSDTAKPHIAAPFKEEEKSPVSAPKLPKAQATSVIRHTADAQLCNHQTCPMKAASILNYQNNSFRRRTHLNVEAARKNIPCAAVSPNRSKCERNTVADVD
RUO
KLF10
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
7071
KLF10 (Human) Recombinant Protein (Q02)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
EGRA/TIEG/TIEG1
KLF10
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers