missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human ITGB7 Partial ORF (NP_000880, 401 a.a. - 505 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00003695-Q01.25ug
This item is not returnable.
View return policy
Description
Sequence: PPGVHISYESQCEGPEKREGKAEDRGQCNHVRINQTVTFWVSLQATHCLPEPHLLRLRALGFSEELIVELHTLCDCNCSDTQPQAPHCSDGQGHLQCGVCSCAPGSpecifications
NP_000880 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.29kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
PPGVHISYESQCEGPEKREGKAEDRGQCNHVRINQTVTFWVSLQATHCLPEPHLLRLRALGFSEELIVELHTLCDCNCSDTQPQAPHCSDGQGHLQCGVCSCAPG | |
RUO | |
ITGB7 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
3695 | |
ITGB7 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ITGB7 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |