missing translation for 'onlineSavingsMsg'
Learn More

Human IL-6, Recombinant Protein, eBioscience™

Recombinant Protein

Brand:  Affymetrix eBioscience 14-8069-80

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15587486

  • £164.35 / 25µg


EU Stock 3
Add to basket

Description

Description

Interleukin 6 is a cytokine secreted by T cells and macrophages, has shown to exhibit activity in response to tissue damage and infection. They have also been found to be involved in various other activities like development of blood, cancer, embryonic development, and regulation of the immune system. IL6 is included in a family of other cytokines which include ciliary neurotrophic factor, cardiotrophin-1, leukemia inhibitory factor and interleukin 11. IL-6 can bring about differentiation through a receptor system which requires a specific receptor and a signaling subunit, gp130. Description: Human IL-6 (also known as B Cell Stimulating Factor BCSF, BSF-2) is a ∽21-28kDa factor produced by a variety of cell types including monocytes, fibroblasts and endothelial cells. Upon stimulation, macrophages, T, B, mast, glial cells, eosinophils, keratinocytes and granulocytes also secrete IL-6. This pleiotrophic cytokine influences antigen-specific immune response, inflammatory response, and the development of neurons and is a major mediator of the acute phase reaction. Applications Reported: Recombinant human IL-6 is biologically active and can promote proliferation of human cell line in culture. It is also used as standard in human IL-6 ELISA. Applications Tested: This recombinant human IL-6 has been tested by sandwich ELISA using anti-human IL-6 antibody pair (MQ2-13A5/MQ2-39C3) and in bioassays using human cell line TF-1. The ED50 measured in a TF-1 proliferation assay is typically less than 0.10ng/mL, corresponding to a specific activity of greater than 1x10e7 Units/mg. Source: E. coli expressed amino acids Pro 29-Met 212 of mature human IL-6 accession # AF372214. Bioactivity: Measured by human TF-1 cell proliferation assay. The ED50 is 0.1ng/mL, corresponding to a specific activity of 1.0 x 10e7 Units/mg. Endotoxin: Less than 0.01ng/ug cytokine as determined by the LAL assay. Purity: >98% as determined by SDS-PAGE. Molecular Weight: The methionylated N-terminal protein has a predicted molecular mass of 21,041. The DTT-reduced and non-reduced protein migrate as 16kDa polypeptides on SDS-PAGE. Storage and handling: For best recovery, quick-spin vial prior to opening. Use in a sterile environment. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2μm post-manufacturing filtered.
Specifications

Specifications

P05231
Cytokine Bioassays
PBS with no preservative; pH 7.2
The methionylated N-terminal protein has a predicted molecular mass of 21,041. The DTT-reduced and non-reduced protein migrate as 16kDa polypeptides on SDS-PAGE.
7.2
Affinity chromatography
E. coli
RUO
Recombinant
Human IL-6
Control, Functional Assay
Human
Hu
Recombinant
0.1mg/mL
Liquid
3569
Human IL-6
SDS-PAGE
25μg
-80°C
E. coli expressed amino acids Pro 29-Met 212 of mature human IL-6 accession # AF372214
Less than 0.01ng/μg cytokine as determined by the LAL assay.
IL6
Measured by human TF-1 cell proliferation assay. The ED50 is 0.1ng/mL, corresponding to a specific activity of 1.0 x 107 Units/mg.
Unlabeled
E. coli
Videos

For Research Use Only.