missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-6 Quantikine HS ELISA Kit, R&D Systems™

a 5.5 hour solid-phase ELISA designed to measure human IL-6 in serum, plasma, and urine.

Brand:  R&D Systems™ HS600B

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13457385

  • £704.13 / 96 tests


Estimated Shipment date
from Supplier 31-03-2020
Add to basket

Description

Description

It contains E. coli-expressed recombinant human IL-6 and has been shown to accurately quantitate the recombinant factor. Results obtained using natural human IL-6 showed linear curves that were parallel to the standard curves obtained using the Quantikine HS kit standards. These results indicate that this kit can be used to determine relative mass values for naturally occurring IL-6
Specifications

Specifications

1 Kit
Human
HRP
ELISA
Solid Phase Sandwich ELISA
0.11pg/mL
Natural and recombinant human IL-6
Urine
0.16 - 10pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
3569
1 Plate
Serum (100 μL), EDTA Plasma (100 μL), Citrate Plasma (100 μL), Urine (100 μL)
Sufficient materials to run ELISAs on at least one 96-well plate
Videos
Documents