missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-6 Quantikine HS ELISA Kit, R&D Systems™

a 5.5 hour solid-phase ELISA designed to measure human IL-6 in serum, plasma, and urine.

Brand:  R&D Systems™ SS600B

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13442876

  • £3506.27 / Pack of 6

Estimated Shipment
01-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

It contains E. coli-expressed recombinant human IL-6 and has been shown to accurately quantitate the recombinant factor. Results obtained using natural human IL-6 showed linear curves that were parallel to the standard curves obtained using the Quantikine HS kit standards. These results indicate that this kit can be used to determine relative mass values for naturally occurring IL-6
Specifications

Specifications

0.16 - 10pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
3569
6 Plates
0.11pg/mL
Serum (100μL), EDTA Plasma (100μL), Citrate Plasma (100μL), Urine (100μL)
Natural and recombinant human IL-6
HRP
ELISA
Solid Phase Sandwich ELISA
1 Pk.
Serum (100uL), EDTA Plasma (100uL), Citrate Plasma (100uL), Urine (100uL)
Human
Sufficient materials to run ELISAs on at least six 96-well plates
Videos