missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-6 QuantiGlo ELISA Kit
Human IL-6 QuantiGlo ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Assay Range | 0.48 to 1500 pg/mL |
---|---|
Assay Sensitivity | 0.35 pg/mL |
Sample Type | Cell Culture Supernatant, Serum, Plasma (EDTA, Citrate, Heparin), Plasma (EDTA, Citrate, Heparin), Serum |
Description
The Human IL-6 QuantiGlo ELISA Kit quantifies human IL-6 in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), citrate plasma (100 ul).Specifications
0.48 to 1500 pg/mL | |
0.35 pg/mL | |
Cell Culture Supernatant, Serum, Plasma (EDTA, Citrate, Heparin), Plasma (EDTA, Citrate, Heparin), Serum |