missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-6 QuantiGlo ELISA Kit, R&D Systems™

Human IL-6 QuantiGlo ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ SQ6000B

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10014953

  • £4348.26 / Pack of 6


Estimated Shipment date
from Supplier 02-04-2020
Add to basket

Description

Description

The Human IL-6 QuantiGlo ELISA Kit quantifies human IL-6 in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), citrate plasma (100 ul).
Specifications

Specifications

1 Pack
Human
HRP
ELISA
Solid Phase Sandwich ELISA
0.35pg/mL
Natural and recombinant human IL-6
Sufficient materials to run ELISAs on at least six 96-well plates
Serum
0.48 - 1500pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
3569
6 Plates
Cell Culture Supernates (100 μL), Serum (100 μL), EDTA Plasma (100 μL), Heparin Plasma (100 μL), Citrate Plasma (100 μL)
Store the unopened product at 2° to 8°C. Do not use past expiration date.
B cell stimulatory factor-2, B-cell differentiation factor, BSF2, BSF-2, BSF2CTL differentiation factor, CDF, HGFHSFIFNB2Hybridoma growth factor, IFNB2, IFN-beta-2, IL6, IL-6B-cell stimulatory factor 2, Interferon beta-2, interleukin 6 (interferon, beta 2), interleukin BSF-2, interleukin-6, MGI-2A
Videos
Documents