missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-5 Quantikine ELISA Kit, R&D Systems™

a 4.5 hour solid phase ELISA designed to measure IL-5 levels in cell culture supernates, serum, plasma, and urine.

Brand:  R&D Systems™ D5000B

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13462565

  • £539.52 / 96 tests


Estimated Shipment
28-02-2019
Add to basket

Description

Description

It contains Sf 21-expressed recombinant human IL-5 and antibodies raised against the recombinant factor and has been shown to accurately quantitate the recombinant factor. Results obtained using natural human IL-5 showed linear curves that were parallel to the standard curves obtained using the Quantikine kit standards. These results indicate that this kit can be used to determine relative mass values for natural IL-5
Specifications

Specifications

3.90 - 250pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
3567
1 Plate
1.08pg/mL
Cell Culture Supernates (100μL), Serum (100μL), EDTA Plasma (100μL), Heparin Plasma (100μL), Urine (100μL)
Natural and recombinant human IL-5
HRP
ELISA
Solid Phase Sandwich ELISA
1 Kit
Cell Culture Supernates (100uL), Serum (100uL), EDTA Plasma (100uL), Heparin Plasma (100uL), Urine (100uL)
Human
Sufficient materials to run ELISAs on at least one 96-well plate
Videos