missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-5 Quantikine ELISA Kit, R&D Systems™
Human IL-5 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Assay Range | 3.90 - 250pg/mL |
---|---|
Assay Sensitivity | 1.08pg/mL |
Sample Type | Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine |
Description
The Human IL-5 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IL-5 in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), urine (100 ul).Specifications
3.90 - 250pg/mL | |
1.08pg/mL | |
Cell Culture Supernatant, Plasma (EDTA, Heparin), Serum, Urine |