missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human IL-5 Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human IL-5.

Brand:  Invitrogen™ BMS278TENCE

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15501877

  • £2175.09 / Pack of 10


Estimated Shipment date
from Supplier 26-02-2018
Add to basket

Disclaimers:

For Research Use Only


Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-5
Biotin-Conjugate anti-human IL-5 monoclonal antibody
Streptavidin-HRP
Human IL-5 Standard lyophilized, 1ng/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

Biotin
10 x 96 tests
Cell culture supernatant, serum
Human
B cell differentiation factor 1, TRF, EDF
ELISA
1.5pg/mL
50μL
IL-5
Videos