missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-4 Quantikine HS ELISA Kit, R&D Systems™

Human IL-4 Quantikine HS ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ HS400

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13447385

  • £721.77 / 96 tests

Estimated Shipment date
from Supplier 17-12-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Human IL-4 Quantikine HS ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IL-4 in cell culture supernates (200 ul), serum (200 ul).
Specifications

Specifications

0.25 - 16pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
3565
1 Plate
0.22pg/mL
Cell Culture Supernates (200μL), Serum (200μL)
Natural and recombinant human IL-4
Sufficient materials to run ELISAs on at least one 96-well plate
HRP
ELISA
Solid Phase Sandwich ELISA
1 Kit
Cell Culture Supernates (200uL), Serum (200uL)
Human
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Videos
Documents