missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-4 Quantikine HS ELISA Kit, R&D Systems™
Human IL-4 Quantikine HS ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Assay Range | 0.25 - 16pg/mL |
---|---|
Assay Sensitivity | 0.22pg/mL |
Sample Type | Cell Culture Supernatant, Serum |
Description
The Human IL-4 Quantikine HS ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IL-4 in cell culture supernates (200 ul), serum (200 ul).Specifications
0.25 - 16pg/mL | |
0.22pg/mL | |
Cell Culture Supernatant, Serum |