missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-4 Quantikine ELISA Kit, R&D Systems™
Human IL-4 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Assay Range | 31.20 - 2000pg/mL |
---|---|
Assay Sensitivity | 10pg/mL |
Sample Type | Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum |
Description
The Human IL-4 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IL-4 in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul), citrate plasma (50 ul).Specifications
31.20 - 2000pg/mL | |
10pg/mL | |
Cell Culture Supernatant, Plasma (EDTA, Citrate, Heparin), Serum |