missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-4 Quantikine ELISA Kit, R&D Systems™

Human IL-4 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ S4050

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13442656

  • £3037.79 / Pack of 6

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Human IL-4 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IL-4 in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul), citrate plasma (50 ul).
Specifications

Specifications

31.20 - 2000pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
3565
6 Plates
10pg/mL
Cell Culture Supernates (50μL), Serum (50μL), EDTA Plasma (50μL), Heparin Plasma (50μL), Citrate Plasma (50μL)
Natural and recombinant human IL-4
Sufficient materials to run ELISAs on at least six 96-well plates
HRP
ELISA
Solid Phase Sandwich ELISA
1 Pk.
Cell Culture Supernates (50uL), Serum (50uL), EDTA Plasma (50uL), Heparin Plasma (50uL), Citrate Plasma (50uL)
Human
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Videos