Human IL-4 ELISA Kits, Thermo Scientific™

Microplate kits for sandwich ELISA of human IL-4 (IL-4)

Overview
Brand: Invitrogen

Manufacturer Part Number: EH3IL45

UNSPSC: 12352203

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 15368224

Quantity Price
1 £ 1330.0 / Pack of 5
Estimated Shipment date
from Supplier 10-04-2017
Add to basket


Description and Specification

Specification

Incubation Time 3.5 hours
Sensitivity ≤ 2pg/mL
Formulation Complete ELISA kit with coated 96-well plates
Sufficient For 5 x 96 microplate wells
Specificity No significant cross-reaction with human IL-2, IL-6, IL-10, IFNγ, GM-CSF or mouse IL-4
Target Natural and recombinant human interleukin-4 (IL-4; also called BSF-1 or BCGF-1)
Precision intra-assay CV ≤ 10%; inter-assay CV ≤ 10%
Sample Size 50μL
Sample Type Cell culture supernatant, serum and plasma (EDTA, sodium citrate and heparin)
Cross Reactivity Human
Quantity 5-plate kit
Includes Anti-Human IL-4 Precoated 96-well Strip Plate, 5 each
Lyophilized Recombinant Human IL-4 Standards, 10 vials
Standard Diluent, 5 x 26mL
30X Wash Buffer, 5 x 50mL
Anti-Human IL-4-HRP Concentrate, 5 x 75μL
HRP Dilution Buffer, 5 x 14mL
TMB Substrate Solution, 5 x 55mL
Stop Solution, 5 x 13mL
Adhesive Plate Covers, 20 each
Test Time 3.5 hours