missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human IL-4 ELISA Kits

Measure levels of human interleukin-4 in human serum, plasma (EDTA, heparin, sodium citrate) and culture supernatant samples using 96-well plates.

Brand:  Invitrogen™ EH3IL4

Code : PN

Additional Details : Weight : 2.49480kg

 View more versions of this product

Product Code. 12301973

  • £464.30 / 1 set


Estimated Shipment
15-06-2018
Add to basket

Description

Description

The human IL-4 ELISA kits use pre-titered, matched pairs of coating and detection antibodies to achieve sensitive and accurate measurement of interleukin-4 (IL-4), also known as B-cell stimulatory factor 1 (BSF-1, BSF1) or B-cell growth factor 1 (BCGF-1, BCGF1), in samples at 50μL per well. Also included are calibrated recombinant protein standard, TMB substrate solution and stop solution. The complete kits contain pre-coated 96-well strip plate(s) and lot-optimized reagents. All components undergo quality control testing to minimize lot-to-lot variability.
  • Target: Natural and recombinant human interleukin-4 (IL-4; also called BSF-1 or BCGF-1)

  • Format: Colorimetric ELISA (TMB substrate) in clear 96 well strip plate

  • Measurement: Absorbance at 450nm minus absorbance at 550nm

  • Assay Range: 10.24 to 400pg/mL

  • Sensitivity: ≤ 2pg/mL

  • Precision: Intra assay CV ≤ 10%; inter-assay CV ≤ 10%

  • Sample Types: Cell culture supernatant, serum and plasma (EDTA, sodium citrate and heparin)

  • Total Assay Time: 3.5 hours

  • Sample Size: 50μL

  • Specificity: No significant cross-reaction with human IL-2, IL-6, IL-10, IFNγ, GM-CSF or mouse IL-4

Includes:Complete kits contain pre-coated antibody-plates, detection antibodies, buffers, diluents, standards, and substrates.

Specifications

Specifications

10.24 to 400pg/mL
Colorimetric ELISA (TMB substrate) in clear 96 well strip plate
TMB Substrate Solution, 55mL, Adhesive Plate Covers, 4 ea, Stop Solution, 13mL, Standard Diluent, 26mL, Lyophilized Recombinant Human IL-4 Standard, 2 vials, HRP Dilution Buffer, 14mL, Anti-Human IL-4 Precoated 96-well Strip Plate, 1 ea, 30X Wash Buffer, 50mL, Anti-Human IL-4-HRP Concentrate, 75µL
Absorbance at 450nm minus absorbance at 550nm
≤ 2pg/mL
50µL
96 microplate wells
Natural and recombinant human interleukin-4 (IL-4; also called BSF-1 or BCGF-1)
Human IL-4 ELISA Kit
Complete ELISA kit with coated 96-well plate
Intra assay CV ≤ 10%; inter-assay CV ≤ 10%
1 plate kit
Cell culture supernatant, serum and plasma (EDTA, sodium citrate and heparin)
No significant cross-reaction with human IL-2, IL-6, IL-10, IFNg, GM-CSF or mouse IL-4
3.5 hr.
Videos