missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-29 (IFN lambda 1) Human Uncoated ELISA Kit with Plates

Specifically engineered for accurate and precise measurement of human IL-29 protein levels from samples including serum, plasma, and supernatants from cell cultures.

Brand:  Invitrogen™ 88-7296-88

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15581097

  • £301.00 / Pack of 10


Estimated Shipment
07-12-2020
Add to basket

Alert:

Contains Azide


Includes: Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant cytokine for generating standard curve and calibrating samples
ELISA/ELISPOT Coating Buffer Powder. This Ready-Set-Go! ELISA Set may contain ELISA/ELISPOT Coating Buffer Powder (Reconstitute to 1L with dH20 and filter (0.22 uM)) or 10X PBS ELISA Coating Buffer (Dilute 1 part 10X Buffer into 9 parts dH20).
Assay Diluent. 5X concentrated
Detection enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Substrate Solution

Description

Description

Human IL-29/IFN-\lambda Ready-SET-Go! ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)
Specifications

Specifications

ELISA
2°C to 8°C except standard which should be stored at ≤-70°C
Interleukin-29
Human
8pg/mL
IL-29 (IFN lambda)
10 x 96 Tests
Videos
SDS
Documents

For Research Use Only