missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human IL-27 ELISA Ready-SET-Go!™ Kit

Allows the specific detection of IL-27 with no detection of IL-35 or free p28 or EBI3. This set has been optimized for the accurate and precise measurement of human IL-27 in serum, plasma, and tissue culture supernatant samples.

Brand:  Invitrogen™ 88-7278-22

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15511097

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Includes: Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant cytokine for generating standard curve and calibrating samples
Coating Buffer. 10X PBS Coating Buffer
Assay Diluent. 5X Concentrated
Detection Enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Solution
Certificate of Analysis. Lot-specific instructions for the dilution of antibodies and standards

Description

Description

Human IL-27 ELISA Ready-SET-Go!™ set includes all of the necessary buffers and reagents to perform enzyme-linked immunosorbent assays. Serum and plasma samples must be diluted 2-fold in prepared assay diluent prior to evaluation in this ELISA.
Specifications

Specifications

ELISA
64pg/mL
2° to 8°C except standard which should be stored at ≤−70°C
Interleukin-27
2 x 96 tests
Human
IL-27
Videos

For Research Use Only