missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Human IL-27 ELISA Ready-SET-Go!™ Kit
Allows the specific detection of IL-27 with no detection of IL-35 or free p28 or EBI3. This set has been optimized for the accurate and precise measurement of human IL-27 in serum, plasma, and tissue culture supernatant samples.
£280.64 - £292.87
Specifications
Assay Sensitivity | 64pg/mL |
---|---|
Target Species | Human |
For Use With (Application) | ELISA |
Quantity | 10 x 96 Tests |
Storage Requirements | 2°C to 8°C except standard which should be stored at ≤-70°C |
Includes: Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant cytokine for generating standard curve and calibrating samples
Coating Buffer. 10X PBS Coating Buffer
Assay Diluent. 5X Concentrated
Detection Enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Solution
Certificate of Analysis. Lot-specific instructions for the dilution of antibodies and standards
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15521097
|
Invitrogen™
88-7278-86 |
10 x 96 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
15531097
|
Invitrogen™
88-7278-88 |
10 x 96 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Human IL-27 ELISA Ready-SET-Go!™ set includes all of the necessary buffers and reagents to perform enzyme-linked immunosorbent assays. Serum and plasma samples must be diluted 2-fold in prepared assay diluent prior to evaluation in this ELISA.Specifications
64pg/mL | |
ELISA | |
2°C to 8°C except standard which should be stored at ≤-70°C | |
IL-27 |
Human | |
10 x 96 Tests | |
Interleukin-27 |
For Research Use Only