missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human IL-27 ELISA Ready-SET-Go!™ Kit

Allows the specific detection of IL-27 with no detection of IL-35 or free p28 or EBI3. This set has been optimized for the accurate and precise measurement of human IL-27 in serum, plasma, and tissue culture supernatant samples.

Brand:  Invitrogen™ 88-7278-86

Code : NEW

Additional Details : Weight : 0.47500kg

 View more versions of this product

Product Code. 15521097

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Includes: Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant cytokine for generating standard curve and calibrating samples
Coating Buffer. 10X PBS Coating Buffer
Assay Diluent. 5X Concentrated
Detection Enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Solution
Certificate of Analysis. Lot-specific instructions for the dilution of antibodies and standards

Description

Description

Human IL-27 ELISA Ready-SET-Go!™ set includes all of the necessary buffers and reagents to perform enzyme-linked immunosorbent assays. Serum and plasma samples must be diluted 2-fold in prepared assay diluent prior to evaluation in this ELISA.
Specifications

Specifications

64pg/mL
ELISA
2°C to 8°C except standard which should be stored at ≤-70°C
IL-27
Human
10 x 96 Tests
Interleukin-27
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only