Learn More
Invitrogen™ IL-21 Human Uncoated ELISA Kit with Plates
Specifically engineered for accurate and precise measurement of IL-21 protein levels from samples including serum, plasma, and cell culture supernatants. This second generation kit has increased sensitivity with a range from 8-1000pg/mL.
Brand: Invitrogen™ 88-8218-76
Code : NEW
Additional Details : Weight : 0.47500kg
Alert:
Contains Sodium azide
Includes: Capture Antibody.Pre-titrated, purified antibody
Detection Antibody.Pre-titrated, biotin-conjugated antibody
Standard.Recombinant protein for generating standard curve and calibrating samples
Coating Buffer.10X PBS ELISA Coating Buffer
Assay Diluent.5X Concentrated
Detection Enzyme.Pre-titrated Avidin-HRP
Substrate Solution.Tetramethylbenzidine (TMB) Solution
Certificate of Analysis. Lot-specific instructions for dilution of antibodies and standards.
Description
Human IL-21 Ready-SET-Go! (2nd Generation) ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)Specifications
8pg/mL | |
ELISA | |
20 x 96 Tests | |
Interleukin-21 |
Human | |
IL-21 | |
2°C to 8°C except standard which should be stored at ≤-70°C | |
IL-21 |
For Research Use Only