missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human IL-2, Recombinant Protein

Human Recombinant Protein

Brand:  Invitrogen™ 14-8029-63

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15527486

  • £107.90 / 10µg


EU Stock 1
Add to basket

Disclaimers:

For Research Use Only.


Description

Description

Human IL-2 (also known as TCGF) is a ∽15 kDa factor produced mainly by activated CD4+ T cells. IL-2 induces cell cycle progression of resting cells in an antigen non-specific manner and allows clonal expansion of activated T cells. IL-2 also acts on activated B cells, monocytes, NK, LAK cells, and on oligodendroglial cells in vitro. In addition, IL-2 plays a role in hematopoiesis, tumor surveillance and anti-inflammatory reactions and hence is a central regulator of the immune response. Non-glycosylated IL-2 is biologically active.
Specifications

Specifications

Cytokine Bioassays
15kDa
SDS-PAGE
10μg
RUO
Less than 0.01ng/μg cytokine as determined by the LAL assay.
FA
Unlabeled
Yes
Sterile liquid: 40mM Acetate, 0.2M NaCl, pH 5, 1.0% BSA . 0.22μm filtered.
IL-2
>98%
E. coli
E. coli
Interleukin-2
The ED50 of this protein, as measured by CTLL-2 proliferation assay, is less than or equal to 175pg/mL. This corresponds to a specific activity of greater than or equal to 5.7 x 106 Units/mg.
Human
Videos