Learn More
Invitrogen™ eBioscience™ Human IL-2 Platinum ELISA Kit, for in vitro diagnostic use
For the quantitative detection of human IL-2.
Brand:  Invitrogen™ BMS221-2TENCE
Code : NEW
Additional Details : Weight : 0.23750kg
 View more versions of this product
Product Code. 15531727
  • £2174.80 / Pack of 10
Estimated Shipment date
from Supplier 26-03-2018
Add to basket
Disclaimers:
For Research Use Only
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-2
Description
Description
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications
Specifications
Biotin
10 x 96 tests
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin), amniotic fluid, urine, whole blood
Human
Aldesleukin, T cell growth factor, TCGF Aldesleukin, T cell growth factor, TCGF
ELISA
9.2pg/mL
50μL
IL-2
Videos