missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ IL-2 Human ELISA Kit
The Human Interleukin-2 (Hu IL-2) ELISA quantitates Hu IL-2 in human serum, plasma or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu IL-2.
£618.00 - £2545.00
Specifications
Accession Number | P60568 |
---|---|
Product Type | ELISA Kit |
Sample Type | Plasma, Serum, Supernatant |
Target Species | Human |
For Use With (Application) | ELISA |
Product Code | Brand | Quantity | Assay Sensitivity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Assay Sensitivity | Price | Quantity & Availability | ||||
15541727
|
Invitrogen™
BMS221-2TWO |
2 x 96 Tests | 9.1 pg/mL |
£618.00
Pack of 2 |
Estimated Shipment: 30-04-2024
Log in to see stock. |
||||
15511727
|
Invitrogen™
BMS221-2FIVE |
5 x 96 Tests | 9.1pg/mL |
£1548.00
Pack of 5 |
Estimated Shipment: 30-04-2024
Log in to see stock. |
||||
15561727
|
Invitrogen™
BMS221-2TEN |
10 x 96 Tests | 9.1pg/mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
Interleukin 2 (IL-2) is an immuno-modulatory cytokine that is important for the proliferation of activated T cells, differentiation of B cells, natural killer cells, monocytes and macrophages. IL-2 signals through the IL-2 receptor (IL-2R), a heterotrimeric protein complex whose gamma chain is also shared by interleukin 4 (IL-4) and interleukin 7 (IL-7). The expression of the IL-2 gene in mature thymocytes is monoallelic, which represents an unusual regulatory mode for controlling the precise expression of a single gene. The targeted disruption of a gene similar to IL-2 in mice leads to an ulcerative colitis-like disease that suggests an essential role of this gene in the immune response to antigenic stimuli.Specifications
P60568 | |
Plasma, Serum, Supernatant | |
ELISA | |
3558 | |
7% | |
RUO | |
2°C to 8°C | |
Human | |
3 hr. 30 min. |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
5% | |
HRP | |
Plasma, 50 μL; Serum, 50 μL; Supernatant, 50 μL | |
Aldesleukin, T cell growth factor, TCGF | |
1 hr. 20 min. |
For Research Use Only