missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-2 DuoSet, R&D Systems™

Human IL-2 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ DY202

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13443205

  • £434.22 / Pack of 15

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Human IL-2 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IL-2.
Specifications

Specifications

ELISA
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
ELISA Development Kit
1 Kit
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Biotin
3558
15 Plates
Human
15.60 - 1000pg/mL
Videos