missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Human IL-1beta Platinum ELISA Kit
For the quantitative detection of human IL-1beta.
Brand: Invitrogen™ BMS224-2
Code : NEW
Additional Details : Weight : 0.23750kg
Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-1beta
Biotin-Conjugate anti-human IL-1beta monoclonal antibody
Streptavidin-HRP
Human IL-1beta Standard lyophilized, 500pg/mL upon reconstitution
Control high, lyophilized
Control low, lyophilized
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films
Description
Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.Specifications
Biotin | |
96 tests | |
Cell culture supernatant, serum, plasma (citrate, heparin), urine | |
Human | |
Catabolin, Interleukin-1 beta, IL1F2 |
ELISA | |
0.3pg/mL | |
50μL | |
IL-1 beta |
For Research Use Only