missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human IL-1beta Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human IL-1beta.

Brand:  Invitrogen™ BMS224-2TENCE

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15581747

  • £2180.00 / Pack of 10


Estimated Shipment date
from Supplier 09-03-2020
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-1beta
Biotin-Conjugate anti-human IL-1beta monoclonal antibody
Streptavidin-HRP
Human IL-1beta Standard lyophilized, 500pg/mL upon reconstitution
Control high, lyophilized
Control low, lyophilized
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

Biotin
0.3pg/mL
IL-1 beta
10 x 96 Tests
Urine
ELISA
50 μL
Catabolin, Interleukin-1 beta, IL1F2
Human
Videos
Documents

For Research Use Only