missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human IL-18 Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human IL-18.

Brand:  Invitrogen™ BMS267-2TENCE

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15531857

  • £2175.09 / Pack of 10


Estimated Shipment date
from Supplier 30-05-2018
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-18
Biotin-Conjugate anti-human IL-18 monoclonal antibody
Streptavidin-HRP
Human IL-18 Standard lyophilized, 10ng/mL upon reconstitution
Control high, lyophilized
Control low, lyophilized
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

Biotin
10 x 96 tests
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Human
Interleukin-18, Interferon-gamma-inducing factor
ELISA
9.0pg/mL
50μL
IL-18
Videos

For Research Use Only