missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ IL-17F (Homodimer) Human Uncoated ELISA Kit

Uncoated ELISA Kit

Brand:  Invitrogen™ 88-7478-88

Code : Z2

Additional Details : Weight : 0.50000kg

Product Code. 13444916

  • £301.00 / 1 set


Estimated Shipment
26-10-2020
Add to basket

Includes: Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant cytokine for generating standard curve and calibrating samples
Coating Buffer. 10X Carbonate ELISA Coating Buffer
Assay Diluent. 5X concentrated
Detection enzyme. Pre-titrated Avidin-HRP
Substrate Solution. Tetramethylbenzidine (TMB) Substrate Solution

Description

Description

IL-17F (Interleukin 17F, CTLA-8)) is a cytokine belonging to the IL-17 family that is produced by inflammatory cells such as activated T cells, mast cells, and basophils. IL-17F is involved in allergic airway inflammation, and can induce several cytokines, chemokines, and adhesion molecules in bronchial epithelial cells, vein endothelial cells, fibroblasts, and eosinophils. IL-17F may be secreted as a homodimer, or a heterodimer with IL17A. It acts by binding to the type I receptor, IL-17R, aiding recruitment of monocytes and neutrophils at the site of inflammation by increasing chemokine production. IL-17F also stimulates induction of other pro-inflammatory cytokines TNF alpha, IL-1 beta, IL-6, and IL-8, and reports strongly suggest the involvement of IL-17 in several chronic inflammatory diseases such as rheumatoid arthritis, psoriasis and multiple sclerosis. TGF-beta (differentiation) and IL-23 (expansion) are required for induction and maintenance of Th17 (IL-17 producing) cells, which in turn induce the other pro-inflammatory cytokines. IL-17F is produced, and exists, as a homo-dimer, with homology to a herpes virus early protein, is one of the six members (IL-17A-F) of this cytokine family, and is well characterized and highly expressed by activated effector memory T cells. IL-17F has been found to inhibit the angiogenesis of endothelial cells and induce endothelial cells to produce IL2, TGFB1/TGFB, and monocyte chemoattractant protein-1.
Specifications

Specifications

Q96PD4
Biotin
112744
ELISA Kit
16 pg/mL
2°C to 8°C
24 hr. 30 min.
IL-2
10 x 96 Tests
Supernatant
Colorimetric Microplate Reader
16-2,000 pg/mL
ELISA
Capture Antibody: Pre-titrated; purified antibody, Detection Antibody: Pre-titrated; biotin-conjugated antibody, Standard: Recombinant cytokine for generating standard curve and calibrating samples, Coating Buffer: 10X Carbonate ELISA Coating Buffer, Assay Diluent: 5X concentrated, Detection enzyme: Pre-titrated Avidin-HRP, Substrate Solution: Tetramethylbenzidine (TMB) Substrate Solution, Certificate of Analysis: Lot-specific instructions for dilution of antibodies and standards, 96 Well Plate: NUNC Maxisorp flat-bottom (included with product Cat. Nos. ending in suffixes -22; -44; -76; -86)
RUO
Plasma, 100 μL|Serum, 100 μL|Supernatant, 100 μL
1 hr. 45 min.
Human
Interleukin-17F, IL-17FF
Human
HRP
Videos
Documents

For Research Use Only