Learn More
Invitrogen™ IL-17A Human ELISA Kit, High Sensitivity
High Sensitivity Sandwich ELISA Kit
Brand: Invitrogen™ BMS2017HS
Code : NEW
Additional Details : Weight : 0.23750kg
Includes: Aluminum pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-17A
Biotin-Conjugate anti-human IL-17A monoclonal antibody
Streptavidin-HRP
Human IL-17A Standard lyophilized, 200pg/mL upon reconstitution
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Amplification Diluent Concentrate (2x)
Amplification Reagent I
Amplification Reagent II
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution
Stop Solution (1M Phosphoric acid)
Adhesive Films
Description
The Human High Sensitivity Interleukin 17A (HS IL-17A) ELISA quantitates Hu HS IL-17A in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu HS IL-17A. Principle of the method The Human HS IL-17A solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. A special amplification reagent step is added to achieve the high sensitivity capability. After incubation and washing steps to rid the microplate of unbound substances, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Interleukin-17A (IL-17A, CTLA-8) is a CD4+ T cell-derived cytokine that promotes inflammatory responses in cell lines and is elevated in rheumatoid arthritis, asthma, multiple sclerosis, psoriasis, and transplant rejection. IL-17A is a 32 kDa long, disulfide-linked homodimer consisting of 136 amino acids that is a member of a six-species family of proteins (IL-17A-17F) and signals through the IL-17 receptor (IL-17R/CDw217). High levels of IL-17A homodimer are produced by activated peripheral blood CD4+ T-cells, and IL-17A also enhances expression of the intracellular adhesion molecule-1 (ICAM-1) in human fibroblasts. In particular, human IL-17A also stimulates epithelial, endothelial, or fibroblastic cells to secrete IL-6, IL-8, G-CSF, and PGE2. In the presence of human IL-17A, fibroblasts can sustain the proliferation of CD34+ hematopoietic progenitors and induce maturation into neutrophils. Mouse, rat, and human IL-17A can induce IL-6 secretion in mouse stromal cells, indicating that all homologs can recognize the mouse receptor. IL-17A regulates the activities of NF-kappa B and mitogen-activated protein kinases, stimulates the expression of IL-6 and cyclooxygenase-2 (PTGS2/COX-2), and enhances the production of nitric oxide (NO).Specifications
Q16552 | |
0.01 pg/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
Ctla-8,Ctla8,IL-17,IL-17A,Il17 | |
8.5% | |
HRP | |
RUO | |
2°C to 8°C | |
Human | |
18 hr. |
0.23-15 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
ELISA | |
3605 | |
6.3% | |
Pre-coated 96 well plate, Standard, Stop Solution, Assay Buffer concentrate, Standard, Sample Diluent, Amplification Reagent I, Amplification Reagent II, Amplification Diluent Concentrate, Wash Buffer, Amplification Reagent II, SAV-HRP, Wash Buffer | |
96 Tests | |
Plasma, 50 μL; Serum, 50 μL; Supernatant, 50 μL | |
Interleukin 17 A, CTLA8 | |
1 hr. 20 min. |
For Research Use Only