Learn More
IL-17A Monoclonal Antibody (3.41.2.6.1), Biotin, Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA1069B
Code : LD
Additional Details : Weight : 0.10000kg
Description
The MA1068 anti-IL17a antibody (clone 3-36.2.7.3) has successfully been paired as the coating antibody in a sandwich ELISA with detection antibody MA1069B (biotinylated conjugate of clone 3-41.2.6.1). Typical dilutions for sandwich ELISA include 1 μg/mL for coating and 0.125 - 0.25 μg/mL for detection.
IL-17A Monoclonal antibody specifically detects IL-17A in Human samples. It is validated for ELISA.Specifications
IL-17A | |
Monoclonal | |
0.5 mg/mL | |
PBS with 4% BSA and proprietary preservative | |
Q16552 | |
IL17A | |
Recominant human IL-17A. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA | |
3.41.2.6.1 | |
Biotin | |
IL17A | |
Ctla8; CTLA-8; cytokine; cytotoxic T-lymphocyte-associated antigen 8; cytotoxic T-lymphocyte-associated protein 8; il 17; Il17; IL-17; IL17A; IL-17a; ILN; Interleukin; interleukin 17; interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8); interleukin 17 precursor; interleukin 17A; interleukin-17; Interleukin17A; interleukin-17A; R-IL-17-A | |
Mouse | |
Protein G | |
RUO | |
3605 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |