missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-17 Quantikine ELISA Kit, R&D Systems™

a 4.5 hour solid phase ELISA designed to measure human IL-17 in cell culture supernates, serum, and plasma.

Brand:  R&D Systems™ D1700

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13462555

  • £539.52 / 96 tests


Estimated Shipment
07-03-2019
Add to basket

Description

Description

It contains E. coli-expressed recombinant human IL-17 and antibodies raised against the recombinant factor. This immunoassay has been shown to quantitate recombinant human IL-17 accurately. Results obtained using natural human IL-17 showed linear curves that were parallel to the standard curves obtained using the recombinant Quantikine kit standards. These results indicate that this kit can be used to determine relative mass values for natural human IL-17
Specifications

Specifications

31.20 - 2000pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
3605
1 Plate
15pg/mL
Cell Culture Supernates (100μL), Serum (100μL), EDTA Plasma (100μL), Heparin Plasma (100μL), Citrate Plasma (100μL)
Natural and recombinant human IL-17
HRP
ELISA
Solid Phase Sandwich ELISA
1 Kit
Cell Culture Supernates (100uL), Serum (100uL), EDTA Plasma (100uL), Heparin Plasma (100uL), Citrate Plasma (100uL)
Human
Sufficient materials to run ELISAs on at least one 96-well plate
Videos