missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-17 Quantikine ELISA Kit, R&D Systems™

Human IL-17 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ S1700

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13452646

  • £3544.17 / Pack of 6


Estimated Shipment date
from Supplier 01-04-2020
Add to basket

Description

Description

The Human IL-17 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IL-17/IL-17A in cell culture supernates (100 ul), serum (100 ul), edta plasma (100 ul), heparin plasma (100 ul), citrate plasma (100 ul).
Specifications

Specifications

1 Pack
Human
HRP
ELISA
Solid Phase Sandwich ELISA
15pg/mL
Natural and recombinant human IL-17
Sufficient materials to run ELISAs on at least six 96-well plates
Serum
31.20 - 2000pg/mL
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
3605
6 Plates
Cell Culture Supernates (100uL),Serum (100uL),EDTA Plasma (100uL),Heparin Plasma (100uL),Citrate Plasma (100uL)
Store the unopened product at 2° to 8°C. Do not use past expiration date.
CTLA8, CTLA-8, CTLA8cytotoxic T-lymphocyte-associated serine esterase 8, Cytotoxic T-lymphocyte-associated antigen 8, IL17, IL17A, IL-17A, IL-17Acytotoxic T-lymphocyte-associated protein 8, IL-17CTLA-8, IL17interleukin-17A, interleukin 17 (cytotoxic T-lymphocyte-associated serine esterase 8), interleukin 17A
Videos
Documents