missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-15 Quantikine ELISA Kit, R&D Systems™

Human IL-15 Quantikine ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ S1500

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13442646

  • £3037.79 / Pack of 6

Estimated Shipment date
from Supplier 27-09-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Human IL-15 Quantikine ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IL-15 in cell culture supernates (50 ul), serum (50 ul), edta plasma (50 ul), heparin plasma (50 ul).
Specifications

Specifications

3.90 - 250pg/mL
Cross-species reactivity not tested. <0.5% cross-reactivity observed with available related molecules.
3600
6 Plates
2pg/mL
Cell Culture Supernates (50μL), Serum (50μL), EDTA Plasma (50μL), Heparin Plasma (50μL)
Store the unopened product at 2 - 8° C. Do not use past expiration date.
HRP
ELISA
Solid Phase Sandwich ELISA
1 Pk.
Cell Culture Supernates (50uL), Serum (50uL), EDTA Plasma (50uL), Heparin Plasma (50uL)
Human
Sufficient materials to run ELISAs on at least six 96-well plates
Videos
Documents