Human IL-12 (total) ELISA Kits, Thermo Scientific™


Overview
Brand: Invitrogen

Manufacturer Part Number: EH2IL12T5

Human IL-15(total) ELISA, 5x96 well plate kit

UNSPSC: 12352203

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 15388224

Quantity Price
1 £ 1384.0 / Pack of 5
Estimated Shipment date
from Supplier 01-02-2017
Add to basket


Description and Specification

Specification

Sensitivity < 5 pg/mL
Measuring Range 25.6 to 1000pg/mL
Specificity No significant cross-reaction with human GM-CSF, IFNα, IFNγ, TNFα, TNFβ, IL-1α, IL-2, IL-3, IL-4, IL-5, IL-6, IL-7, IL-8, IL-9, IL-10, IL-11, IL-13, IL-15, mouse IL-12 (p70) or mouse IL-12 (p40).
Sample Type Cell culture supernatant, serum, plasma (EDTA, sodium citrate and heparin)
Sample Size 50μL
Target Total (p70 and p40) natural and recombinant human interleukin-12 (IL-12)
Precision Intra-assay CV < 10%; inter-assay CV < 10%
Test Time 3 hours