missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Mouse CD254 (RANK Ligand), Recombinant Protein

Mouse Recombinant Protein

Brand:  Invitrogen™ 14-8612-62

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15557546

  • £87.42 / 5µg


Estimated Shipment date
from Supplier 27-06-2018
Add to basket

Description

Description

RANK Ligand (Receptor Activator for Nuclear Factor κB Ligand), also known as TNF-related activation-induced cytokine (TRANCE), along with RANK are members of the TNF superfamily. RANK-L can be found as a soluble protein expressed by activated T lymphocytes, which plays a role in dendritic cell maturation. In addition it has also been shown to be cleaved by TACE (TNF-α convertase) releasing an ectodomain variant of the cell bound form. Lastly RANK-L is expressed as a Type II transmembrane protein expressed by osteoblast lineages. The receptor RANK, located on osteoclastic and dendritic cells can recognize all forms of RANK-L. RANK-L plays a critical role in bone development and mineralization. High levels of RANK-L have been associated with degenerative bone diseases.
Specifications

Specifications

0.1mg/mL
Sterile liquid; phosphate buffered saline, pH 7.2, 1.0% BSA. 0.22μm filtered.
CD254 (RANK Ligand)
>90%
E. coli
RUO
Less than 0.01ng/μg cytokine as determined by the LAL assay.
The ED50, as measured by induction of osteoclast formation on mouse RAW264.7 cells, is 4.8ng/mL.
Unlabeled
Yes
Cytokine Bioassays
20kDa
reducing SDS-PAGE
5μg
Store at less than or equal to -70°C.
E. coli expressed amino acid Lys158 - Asp316 Accession # Q3TWY5
RANKL, TRANCE, Receptor Activator for Nuclear Factor kappa B Ligand, Osteoprotegerin ligand, OPGL, ODF, Osteoclast Differentiation Factor
Recombinant Protein
Mouse
Videos