missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human IL-12 p70 Quantikine HS ELISA Kit, R&D Systems™

Human IL-12 p70 Quantikine HS ELISA Kit from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.

Brand:  R&D Systems™ HS120

Code : Z1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13437385

  • £721.77 / 96 tests

Estimated Shipment date
from Supplier 19-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The Human IL-12 p70 Quantikine HS ELISA Kit is a Solid Phase Sandwich ELISA that quantifies human IL-12 p70/IL-12A in cell culture supernates (200 ul), serum (200 ul), edta plasma (200 ul), heparin plasma (200 ul).
Specifications

Specifications

0.63 - 40pg/mL (Cell Culture Supernates), 0.78 - 50pg/mL (Serum, Heparin Plasma, EDTA Plasma)
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules.
3592
1 Plate
0.5pg/mL
Cell Culture Supernates (200μL), Serum (200μL), EDTA Plasma (200μL), Heparin Plasma (200μL)
Natural and recombinant human IL-12p70
Sufficient materials to run ELISAs on at least one 96-well plate
HRP
ELISA
Solid Phase Sandwich ELISA
1 Kit
Cell Culture Supernates (200uL), Serum (200uL), EDTA Plasma (200uL), Heparin Plasma (200uL)
Human
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Videos
Documents