missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human IL-12 (p70) Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human IL-12 p70.

Brand:  Invitrogen™ BMS238CE

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15531797

  • £361.14 / 96 tests


EU Stock 4
Add to basket

Disclaimers:

For Research Use Only


Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-12 p70
Biotin-Conjugate anti-human IL-12 p70 monoclonal antibody
Streptavidin-HRP
Human IL-12 p70 Standard lyophilized, 400pg/mL upon reconstitution
Control high, lyophilized
Control low, lyophilized
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

Biotin
96 tests
Cell culture supernatant, serum, plasma (EDTA, citrate, heparin)
Human
Interleukin-12 subunit p70, CLMF, NKSF
ELISA
2.1pg/mL
50μL
IL-12
Videos