missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Human IL-12 (p70) Platinum ELISA Kit, for in vitro diagnostic use

For the quantitative detection of human IL-12 p70.

Brand:  Invitrogen™ BMS238TENCE

Code : NEW

Additional Details : Weight : 0.23750kg

 View more versions of this product

Product Code. 15561797

  • £2180.00 / Pack of 10


Estimated Shipment date
from Supplier 10-03-2020
Add to basket

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human IL-12 p70
Biotin-Conjugate anti-human IL-12 p70 monoclonal antibody
Streptavidin-HRP
Human IL-12 p70 Standard lyophilized, 400pg/mL upon reconstitution
Control high, lyophilized
Control low, lyophilized
Sample Diluent
Assay Buffer Concentrate 20x (PBS with 1% Tween 20 and 10% BSA)
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Red-Dye
Adhesive Films

Description

Description

Platinum Kits undergo stringent in-house development, manufacture and quality-validation processes ensuring highest possible product performance and optimal lot-to-lot consistency.
Specifications

Specifications

Biotin
2.1pg/mL
IL-12
10 x 96 Tests
Serum
ELISA
50 μL
Interleukin-12 subunit p70, CLMF, NKSF
Human
Videos
Documents

For Research Use Only