missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human IL-12 p70 DuoSet, R&D Systems™
Human IL-12 p70 DuoSet ELISA from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
POA
Specifications
Assay Range | 31.20 to 2000 pg/mL |
---|---|
Cross Reactivity | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. |
Gene ID (Entrez) | 3592 |
Product Type | ELISA Development Kit |
Conjugate | Biotin |
Description
The Human IL-12 p70 DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human IL-12 p70/IL-12A.Specifications
31.20 to 2000 pg/mL | |
3592 | |
Biotin |
<50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules. | |
ELISA Development Kit | |
Human |