Learn More
Invitrogen™ Human IL-10 ProQuantum Immunoassay Kit
ProQuantum Immunoassay Kit
Brand: Invitrogen™ A35590
432.02 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Additional Details : Weight : 0.22720kg
Description
Human IL-10 ProQuantum Immunoassay Kit
ProQuantum Immunoassay Kit
Specifications
Contains assay dilution buffer, antibody-conjugate A, antibody-conjugate B, antibody-conjugate dilution buffer, standard, master mix, and ligase. Store at -5°C to -30°C. | |
96 Tests |