missing translation for 'onlineSavingsMsg'
Learn More

eBioscience™ Mouse TNF alpha, Recombinant Protein, Carrier-Free

Mouse Recombinant Protein

Brand:  Affymetrix eBioscience 34-8321-82

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15508786

  • £324.73 / 100µg


Estimated Shipment date
from Supplier 26-02-2018
Add to basket

Description

Description

Mouse TNF alpha is a 17 kDa factor produced by macrophages, monocytes, neutrophils, CD4+ T cells and NK cells. A 26 kDa form of TNF alpha is expressed as a membrane-bound molecule. TNF alpha is cytolytic and plays an important role in immune regulation. Dimers and trimers of TNF alpha have been observed.
Specifications

Specifications

NM_013693
Cytokine Bioassays
The protein is not methionylated at the N-terminal and has a predicted molecular mass of 17,258. The DTT-reduced and non-reduced protein migrate as 17kDa polypeptides on SDS-PAGE.
SDS-PAGE
100μg
Store at less than or equal to -70°C.
E. coli expressed amino acids Leu 80-Leu 235 of mouse TNF alpha accession # NM_013693
Tumor Necrosis Factor alpha, TNFa TNF-a
Recombinant Protein, Carrier-Free
Mouse
0.5mg/mL
Sterile liquid; 10mM sodium phosphate, pH 7.2, 0.22μm filtered.
TNF alpha
>98%
E. coli
RUO
Less than 0.01ng/μg cytokine as determined by the LAL assay.
The ED50 of this protein, as measured by L929 cytotoxicity assay, is less than or equal to 25pg/mL. This corresponds to a specific activity of greater than or equal to 4 x 107 Units/mg.
Unlabeled
Yes
Videos