missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ eBioscience™ Mouse Flt3 Ligand, Recombinant Protein

Mouse Recombinant Protein

Brand:  Invitrogen™ 14-8001-62

Code : NEW

Additional Details : Weight : 0.09500kg

 View more versions of this product

Product Code. 15567476

  • £56.64 / 5µg


Estimated Shipment date
from Supplier 26-02-2018
Add to basket

Description

Description

Flt3 (FMS-related tyrosine kinase 3) Ligand is a growth factor that stimulates the growth and differentiation of early hematopoetic progenitor cells. Flt3 Ligand alone exerts weak biological effects, and instead synergizes with other colony-stimulating factors to induce the proliferation and differentiation of cells of the myeloid and lymphoid lineages. Recent studies have suggested that Flt3 Ligand plays an important role in the development and expansion of dendritic cells in vivo, and that it may influence NK cell survival by this mechanism.

Flt3 Ligand transcripts are detectable in most cell types and tissues, although the protein has only been found in stromal cells and T cells. It is expressed most often as a transmembrane cell surface protein, although processing can yield a soluble, secreted isoform.
Specifications

Specifications

NM_013520
Cytokine Bioassays
20.7kDa
SDS-PAGE
5μg
Store at less than or equal to -70°C.
E. coli expressed amino acids Thr28-Gln189 accession number AL2
FL, FLT-3L, FMS-related tyrosine kinase 3 Ligand, Flk-2 Ligand
Recombinant Protein
Mouse
0.1mg/mL
Sterile liquid; 20mM glycine, 150mM NaCl, 1% BSA, pH 3.0
Flt3 Ligand
>97%
E. coli
RUO
Less than 0.05ng/μg cytokine as determined by the LAL assay.
The ED50 of this protein, as measured by IL-6 induction in M1 cells, is 50-250ng/mL in the presence of 1ng/mL Mouse LIF Recombinant Protein. This corresponds to a specific activity of 2 x 104 - 4 x 103 Units/mg.
Unlabeled
Yes
Videos