Learn More
Invitrogen™ eBioscience™ Human IL-1 beta ELISA Ready-SET-Go!™ Kit, 2nd Generation
Engineered for accurate measurement of IL-1 levels from samples including serum, plasma, and cell culture supernatants. It recognizes the cleaved mature form and uncleaved pro-form of Human IL-1 beta. This second generation kit has increased sensitivity with a range from 2 to 150pg/mL.
Brand: Invitrogen™ 88-7261-22
Code : NEW
Additional Details : Weight : 0.47500kg
Includes: Capture Antibody: Pre-titrated, purified antibody
Detection Antibody. Pre-titrated, biotin-conjugated antibody
Standard. Recombinant protein for generating standard curve and calibrating samples
10X Coating Buffer . Buffer for plating the Capture Antibody
5X ELISA/ELISPOT Diluent . Buffer for blocking and diluting the Detection Antibody and Enzyme
Enzyme. Pre-titrated Avidin-HRP
Substrate. 1X TMB Solution
Description
Human IL-1 beta Ready-SET-Go! (2nd Generation) ELISA Set contains the necessary reagents, standards, buffers and diluents for performing quantitative enzyme-linked immunosorbent assays (ELISA)Specifications
ELISA | |
2 x 96 tests | |
Human | |
IL-1 beta |
96-well plates | |
2pg/mL | |
2° to 8°C | |
Interleukin-1 beta |
For Research Use Only